Merck
  • Targeted degradation of aberrant tau in frontotemporal dementia patient-derived neuronal cell models.
Targeted degradation of aberrant tau in frontotemporal dementia patient-derived neuronal cell models.
eLife (2019-03-26)
M Catarina Silva, Fleur M Ferguson, Quan Cai, Katherine A Donovan, Ghata Nandi, Debasis Patnaik, Tinghu Zhang, Hai-Tsang Huang, Diane E Lucente, Bradford C Dickerson, Timothy J Mitchison, Eric S Fischer, Nathanael S Gray, Stephen J Haggarty
ABSTRACT
Tauopathies are neurodegenerative diseases characterized by aberrant forms of tau protein accumulation leading to neuronal death in focal brain areas. Positron emission tomography (PET) tracers that bind to pathological tau are used in diagnosis, but there are no current therapies to eliminate these tau species. We employed targeted protein degradation technology to convert a tau PET-probe into a functional degrader of pathogenic tau. The hetero-bifunctional molecule QC-01-175 was designed to engage both tau and Cereblon (CRBN), a substrate-receptor for the E3-ubiquitin ligase CRL4CRBN, to trigger tau ubiquitination and proteasomal degradation. QC-01-175 effected clearance of tau in frontotemporal dementia (FTD) patient-derived neuronal cell models, with minimal effect on tau from neurons of healthy controls, indicating specificity for disease-relevant forms. QC-01-175 also rescued stress vulnerability in FTD neurons, phenocopying CRISPR-mediated MAPT-knockout. This work demonstrates that aberrant tau in FTD patient-derived neurons is amenable to targeted degradation, representing an important advance for therapeutics.
Sigma-Aldrich
Pomalidomide, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Pricing and availability is not currently available.
Sigma-Aldrich
Lenalidomide, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-MAP2 Antibody, Chemicon®, from chicken
Pricing and availability is not currently available.
Sigma-Aldrich
trans-2-Phenylcyclopropylamine hydrochloride
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Pricing and availability is not currently available.