Merck
  • Whole-Exome Enrichment with the Roche NimbleGen SeqCap EZ Exome Library SR Platform.
Whole-Exome Enrichment with the Roche NimbleGen SeqCap EZ Exome Library SR Platform.
Cold Spring Harbor protocols (2015-03-13)
Rui Chen, Hogune Im, Michael Snyder
ABSTRACT
Multiple platforms are available for whole-exome enrichment and sequencing (WES). This protocol is based on the Roche NimbleGen SeqCap EZ Exome Library SR platform, which enriches for ∼44 Mb of the human exonic regions. The SeqCap system uses 55- to 105-base DNA probes to capture known coding DNA sequences (CDS) from the NCBI Consensus CDS Database, RefSeq, and Sanger miRBase. The protocol can be performed at the benchside without the need for automation, and the resulting library can be used for targeted next-generation sequencing on an Illumina HiSeq 2000 sequencer.
Sigma-Aldrich
Anti-Eck/EphA2 Antibody, clone D7, clone D7, Upstate®, from mouse