Merck
  • Merozoite surface protein-1 of Plasmodium yoelii fused via an oligosaccharide moiety of cholera toxin B subunit glycoprotein expressed in yeast induced protective immunity against lethal malaria infection in mice.
Merozoite surface protein-1 of Plasmodium yoelii fused via an oligosaccharide moiety of cholera toxin B subunit glycoprotein expressed in yeast induced protective immunity against lethal malaria infection in mice.
Vaccine (2011-11-29)
Takeshi Miyata, Tetsuya Harakuni, Toki Taira, Goro Matsuzaki, Takeshi Arakawa
ABSTRACT
Methylotrophic yeast (Pichia pastoris) secreted cholera toxin B subunit (CTB) predominantly as a biologically active pentamer (PpCTB) with identical ganglioside binding affinity profiles to that of choleragenoid. Unlike choleragenoid, however, the PpCTB did not induce a footpad edema response in mice. Of the two potential glycosylation sites (NIT(4-6) and NKT(90-92)) for this protein, a N-linked oligosaccharide was identified at Asn4. The oligosaccharide, presumed to extend from the lateral circumference of the CTB pentamer ring structure, was exploited as a site-specific anchoring scaffold for the C-terminal 19-kDa merozoite surface protein-1 (MSP1-19) of the rodent malaria parasite, Plasmodium yoelii. Conjugation of MSP1-19 to PpCTB via its oligosaccharide moiety induced higher protective efficacy against lethal parasite infection than conjugation directly to the PpCTB protein body in both intranasal and subcutaneous immunization regimes. Such increased protection was potentially due to the higher antigen loading capacity of CTB achieved when the antigen was linked to the extended branches of the oligosaccharide. This might have allowed the antigen to reside in more spacious molecular environment with less steric hindrance between the constituent molecules of the fusion complex.
Sigma-Aldrich
Monosialoganglioside GM3 from canine blood, ≥98%, lyophilized powder
Sigma-Aldrich
Disialoganglioside-GD2 from bovine brain, ~95%, lyophilized powder, semisynthetic