Merck
  • Microtubule nucleation during central spindle assembly requires NEDD1 phosphorylation on serine 405 by Aurora A.
Microtubule nucleation during central spindle assembly requires NEDD1 phosphorylation on serine 405 by Aurora A.
Journal of cell science (2019-04-28)
Thibault Courthéoux, David Reboutier, Thibaut Vazeille, Jean-Yves Cremet, Christelle Benaud, Isabelle Vernos, Claude Prigent
ABSTRACT
During mitosis, the cell sequentially constructs two microtubule-based spindles to ensure faithful segregation of chromosomes. A bipolar spindle first pulls apart the sister chromatids, then a central spindle further separates them away. Although the assembly of the first spindle is well described, the assembly of the second remains poorly understood. We report here that the inhibition of Aurora A leads to an absence of the central spindle resulting from a lack of nucleation of microtubules in the midzone. In the absence of Aurora A, the HURP (also known as DLGAP5) and NEDD1 proteins that are involved in nucleation of microtubules fail to concentrate in the midzone. HURP is an effector of RanGTP, whereas NEDD1 serves as an anchor for the γ-tubulin ring complex (γTURC). Interestingly, Aurora A phosphorylates HURP and NEDD1 during assembly of the initial bipolar spindle. We show here that the expression of a NEDD1 isoform mimicking phosphorylation by Aurora A is sufficient to restore microtubule nucleation in the midzone under conditions of Aurora A inhibition. These results reveal a new control mechanism of microtubule nucleation by Aurora A during assembly of the central spindle.