Merck
CERKL regulates autophagy via the NAD-dependent deacetylase SIRT1.
Autophagy (2018-09-13)
Xuebin Hu, Zhaojing Lu, Shanshan Yu, James Reilly, Fei Liu, Danna Jia, Yayun Qin, Shanshan Han, Xiliang Liu, Zhen Qu, Yuexia Lv, Jingzhen Li, Yuwen Huang, Tao Jiang, Haibo Jia, Qing Wang, Jingyu Liu, Xinhua Shu, Zhaohui Tang, Mugen Liu
ABSTRACT
Macroautophagy/autophagy is an important intracellular mechanism for the maintenance of cellular homeostasis. Here we show that the CERKL (ceramide kinase like) gene, a retinal degeneration (RD) pathogenic gene, plays a critical role in regulating autophagy by stabilizing SIRT1. In vitro and in vivo, suppressing CERKL results in impaired autophagy. SIRT1 is one of the main regulators of acetylation/deacetylation in autophagy. In CERKL-depleted retinas and cells, SIRT1 is downregulated. ATG5 and ATG7, 2 essential components of autophagy, show a higher degree of acetylation in CERKL-depleted cells. Overexpression of SIRT1 rescues autophagy in CERKL-depleted cells, whereas CERKL loses its function of regulating autophagy in SIRT1-depleted cells, and overexpression of CERKL upregulates SIRT1. Finally, we show that CERKL directly interacts with SIRT1, and may regulate its phosphorylation at Ser27 to stabilize SIRT1. These results show that CERKL is an important regulator of autophagy and it plays this role by stabilizing the deacetylase SIRT1.
Sigma-Aldrich
Rapamycin, ≥95% (HPLC), powder
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Bafilomycin A1, Streptomyces griseus, Bafilomycin A1, CAS 88899-55-2, acts as a highly potent and specific inhibitor of vacuolar-type H+-ATPase (Ki = 500 pM). Blocks the fusion of autophagosome with lysosome.