Merck
  • CCDC102B functions in centrosome linker assembly and centrosome cohesion.
CCDC102B functions in centrosome linker assembly and centrosome cohesion.
Journal of cell science (2018-11-09)
Yuqing Xia, Ning Huang, Zhiquan Chen, Fangyuan Li, Guiliang Fan, Dandan Ma, Jianguo Chen, Junlin Teng
ABSTRACT
The proteinaceous centrosome linker is an important structure that allows the centrosome to function as a single microtubule-organizing center (MTOC) in interphase cells. However, the assembly mechanism of the centrosome linker components remains largely unknown. In this study, we identify CCDC102B as a new centrosome linker protein that is required for maintaining centrosome cohesion. CCDC102B is recruited to the centrosome by C-Nap1 (also known as CEP250) and interacts with the centrosome linker components rootletin and LRRC45. CCDC102B decorates and facilitates the formation of rootletin filaments. Furthermore, CCDC102B is phosphorylated by Nek2A (an isoform encoded by NEK2) and is disassociated from the centrosome at the onset of mitosis. Together, our findings reveal a molecular role for CCDC102B in centrosome cohesion and centrosome linker assembly.This article has an associated First Person interview with the first authors of the paper.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Pricing and availability is not currently available.