Merck
  • 11β-Hydroxysteroid Dehydrogenase 1 Human Tissue Distribution, Selective Inhibitor, and Role in Doxorubicin Metabolism.
11β-Hydroxysteroid Dehydrogenase 1 Human Tissue Distribution, Selective Inhibitor, and Role in Doxorubicin Metabolism.
Drug metabolism and disposition: the biological fate of chemicals (2018-04-21)
Xin Yang, Wenyi Hua, Sangwoo Ryu, Phillip Yates, Cheng Chang, Hui Zhang, Li Di
ABSTRACT
11β-Hydroxysteroid dehydrogenase 1 (11β-HSD1) is distributed mainly in the human liver, with no detectable levels in the intestine or kidney, based on a newly developed proteomic approach. 11β-HSD1 is mostly membrane-bound and retained in the liver microsomal fraction. Interindividual variability of 11β-HSD1 is relatively low, with about a 3-fold difference. A significant correlation was not observed between various demographic variables (ethnicity, gender, age, weight, smoking, and alcohol use) and 11β-HSD1 protein expression or activity based on data from 31 donors. PF-915275 has been identified as a selective 11β-HSD1 inhibitor with minimal effects on carbonyl reductase 1 and major cytochrome P450 enzymes. 11β-HSD1 has been shown, for the first time, to be involved in doxorubicin metabolism, accounting for approximately 30% of doxorubicinol formation in human hepatocytes.
Sigma-Aldrich
PF-915275, ≥98% (HPLC)