Merck
  • PD-1 Expression during Acute Infection Is Repressed through an LSD1-Blimp-1 Axis.
PD-1 Expression during Acute Infection Is Repressed through an LSD1-Blimp-1 Axis.
Journal of immunology (Baltimore, Md. : 1950) (2019-12-08)
Alexander P R Bally, Dennis K Neeld, Peiyuan Lu, Parimal Majumder, Yan Tang, Benjamin G Barwick, Qing Wang, Jeremy M Boss
ABSTRACT
During prolonged exposure to Ags, such as chronic viral infections, sustained TCR signaling can result in T cell exhaustion mediated in part by expression of programmed cell death-1 (PD-1) encoded by the Pdcd1 gene. In this study, dynamic changes in histone H3K4 modifications at the Pdcd1 locus during ex vivo and in vivo activation of CD8 T cells suggested a potential role for the histone H3 lysine 4 demethylase LSD1 in regulating PD-1 expression. CD8 T cells lacking LSD1 expressed higher levels of Pdcd1 mRNA following ex vivo stimulation as well as increased surface levels of PD-1 during acute, but not chronic, infection with lymphocytic choriomeningitis virus (LCMV). Blimp-1, a known repressor of PD-1, recruited LSD1 to the Pdcd1 gene during acute, but not chronic, LCMV infection. Loss of DNA methylation at Pdcd1's promoter-proximal regulatory regions is highly correlated with its expression. However, following acute LCMV infection, in which PD-1 expression levels return to near baseline, LSD1-deficient CD8 T cells failed to remethylate the Pdcd1 locus to the levels of wild-type cells. Finally, in a murine melanoma model, the frequency of PD-1-expressing tumor-infiltrating LSD1-deficient CD8 T cells was greater than in wild type. Thus, LSD1 is recruited to the Pdcd1 locus by Blimp-1, downregulates PD-1 expression by facilitating the removal of activating histone marks, and is important for remethylation of the locus. Together, these data provide insight into the complex regulatory mechanisms governing T cell immunity and regulation of a critical T cell checkpoint gene.
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys27) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-monomethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.