Merck
  • Pituitary tumour fibroblast-derived cytokines influence tumour aggressiveness.
Pituitary tumour fibroblast-derived cytokines influence tumour aggressiveness.
Endocrine-related cancer (2019-10-24)
Pedro Marques, Sayka Barry, Eivind Carlsen, David Collier, Amy Ronaldson, Sherine Awad, Neil Dorward, Joan Grieve, Nigel Mendoza, Samiul Muquit, Ashley B Grossman, Frances Balkwill, Márta Korbonits
ABSTRACT
Tumour-associated fibroblasts (TAFs) are key elements of the tumour microenvironment, but their role in pituitary neuroendocrine tumours (PitNETs) has been little explored. We hypothesised that TAF-derived cytokines may play a role in tumour aggressiveness and that their release can be inhibited by somatostatin analogues. TAFs were isolated and cultured from 16 PitNETs (11 clinically non-functioning tumours and 5 somatotropinomas). The fibroblast secretome was assessed with a 42-plex cytokine array before and after multiligand somatostatin receptor agonist pasireotide treatment. Angiogenesis and epithelial-to-mesenchymal transition pathway assessment included CD31, E-cadherin and ZEB1 expression. GH3 cells treated with TAF- or skin fibroblast-conditioned medium were assessed for migration, invasion and cell morphology changes. PitNET TAFs secreted significant amounts of cytokines including CCL2, CCL11, VEGF-A, CCL22, IL-6, FGF-2 and IL-8. TAFs from PitNETs with cavernous sinus invasion secreted higher IL-6 levels compared to fibroblasts from non-invasive tumours (P = 0.027). Higher CCL2 release from TAFs correlated with more capillaries (r = 0.672, P = 0.004), and TAFs from PitNETs with a higher Ki-67 tended to secrete more CCL2 (P = 0.058). SST1 is the predominant somatostatin receptor in TAFs, and pasireotide decreased TAF-derived IL-6 by 80% (P < 0.001) and CCL2 by 35% (P = 0.038). GH3 cells treated with TAF-conditioned medium showed increased migration and invasion compared to cells treated with skin fibroblast-conditioned medium, with morphological and E-cadherin and ZEB1 expression changes suggesting epithelial-to-mesenchymal transition. TAF-derived cytokines may increase PitNET aggressiveness, alter angiogenesis and induce epithelial-to-mesenchymal transition changes. Pasireotide's inhibitory effect on TAF-derived cytokines suggest that this effect may play a role in its anti-tumour effects.
Sigma-Aldrich
Gentamicin solution, 50 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Giemsa stain, certified by the Biological Stain Commission
Sigma-Aldrich
Fluoroshield with DAPI, liquid
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture