Merck
  • Glycosylation of Specific Notch EGF Repeats by O-Fut1 and Fringe Regulates Notch Signaling in Drosophila.
Glycosylation of Specific Notch EGF Repeats by O-Fut1 and Fringe Regulates Notch Signaling in Drosophila.
Cell reports (2019-11-14)
Ashutosh Pandey, Beth M Harvey, Mario F Lopez, Atsuko Ito, Robert S Haltiwanger, Hamed Jafar-Nejad
ABSTRACT
Fringe glycosyltransferases differentially modulate the binding of Notch receptors to Delta/DLL versus Serrate/Jagged ligands by adding GlcNAc to O-linked fucose on Notch epidermal growth factor-like (EGF) repeats. Although Notch has 22 O-fucosylation sites, the biologically relevant sites affecting Notch activity during animal development in vivo in the presence or absence of Fringe are not known. Using a variety of assays, we find important roles in Drosophila Notch signaling for GlcNAc-fucose-O glycans on three sites: EGF8, EGF9, and EGF12. O-Fucose monosaccharide on EGF12 (in the absence of Fringe) is essential for Delta-mediated lateral inhibition in embryos. However, wing vein development depends on the addition of GlcNAc to EGF8 and EGF12 by Fringe, with a minor contribution from EGF9. Fringe modifications of EGF8 and EGF12 together prevent Notch from cis-inhibiting Serrate, thereby promoting normal wing margin formation. Our work shows the combinatorial and context-dependent roles of GlcNAc-fucose-O glycans on these sites in Drosophila Notch-ligand interactions.
Sigma-Aldrich
Methotrexate, meets USP testing specifications