Merck
  • Clozapine and other 5-hydroxytryptamine-2A receptor antagonists alter the subcellular distribution of 5-hydroxytryptamine-2A receptors in vitro and in vivo.
Clozapine and other 5-hydroxytryptamine-2A receptor antagonists alter the subcellular distribution of 5-hydroxytryptamine-2A receptors in vitro and in vivo.
Neuroscience (1999-06-12)
D L Willins, S A Berry, L Alsayegh, J R Backstrom, E Sanders-Bush, L Friedman, B L Roth
ABSTRACT
In this study, we demonstrate that clozapine and other atypical antipsychotic drugs induce a paradoxical internalization of 5-hydroxytryptamine-2A receptors in vitro and a redistribution of 5-hydroxytryptamine-2A receptors in vivo. We discovered that clozapine, olanzapine, risperidone and the putative atypical antipsychotic drug MDL 100,907 all induced 5-hydroxytryptamine-2A receptor internalization in fibroblasts stably expressing the 5-hydroxytryptamine-2A receptor in vitro. Two 5-hydroxytryptamine-2A antagonists (mianserin and ritanserin), which have been demonstrated to reduce negative symptoms in schizophrenia, also caused 5-hydroxytryptamine-2A receptor internalization. Four different drugs, each devoid of 5-hydroxytryptamine-2A antagonist activity, had no effect on the subcellular distribution of 5-hydroxytryptamine-2A receptors in vitro. Treatment of rats for seven days with clozapine induced an increase in intracellular 5-hydroxytryptamine-2A receptor-like immunoreactivity in pyramidal neurons, while causing a decrease in labeling of apical dendrites in the medial prefrontal cortex. This redistribution of 5-hydroxytryptamine-2A receptors in pyramidal neurons was also seen when rats were chronically treated with another atypical antipsychotic drug, olanzapine. The typical antipsychotic drug haloperidol, however, did not induce a redistribution of 5-hydroxytryptamine-2A receptors in pyramidal neurons in the medial prefrontal cortex. Taken together, these results demonstrate that several atypical antipsychotic drugs with high 5-hydroxytryptamine-2A receptor affinities induce a redistribution of 5-hydroxytryptamine-2A receptors both in vivo and in vitro. It is conceivable that the loss of 5-hydroxytryptamine-2A receptors from the apical dendrites of pyramidal neurons is important for the beneficial effects of atypical antipsychotic drugs and other 5-hydroxytryptamine-2A antagonists in schizophrenia.