Merck
Nanoscale coupling of endocytic pit growth and stability.
Science advances (2019-12-07)
Martin Lehmann, Ilya Lukonin, Frank Noé, Jan Schmoranzer, Cecilia Clementi, Dinah Loerke, Volker Haucke
ABSTRACT
Clathrin-mediated endocytosis, an essential process for plasma membrane homeostasis and cell signaling, is characterized by stunning heterogeneity in the size and lifetime of clathrin-coated endocytic pits (CCPs). If and how CCP growth and lifetime are coupled and how this relates to their physiological function are unknown. We combine computational modeling, automated tracking of CCP dynamics, electron microscopy, and functional rescue experiments to demonstrate that CCP growth and lifetime are closely correlated and mechanistically linked by the early-acting endocytic F-BAR protein FCHo2. FCHo2 assembles at the rim of CCPs to control CCP growth and lifetime by coupling the invagination of early endocytic intermediates to clathrin lattice assembly. Our data suggest a mechanism for the nanoscale control of CCP growth and stability that may similarly apply to other metastable structures in cells.
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Cysteamine, ≥98.0% (RT)
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, ≥95% (TLC), liquid
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)