Merck
  • Dephosphorylation activates the interferon-stimulated Schlafen family member 11 in the DNA damage response.
Dephosphorylation activates the interferon-stimulated Schlafen family member 11 in the DNA damage response.
The Journal of biological chemistry (2019-08-10)
Dane Malone, Rea M Lardelli, Manqing Li, Michael David
ABSTRACT
Human Schlafen 11 (SLFN11) is an interferon-stimulated gene (ISG) that we previously have demonstrated to ablate translation of HIV proteins based on the virus's distinct codon preference. Additionally, lack of SLFN11 expression has been linked to the resistance of cancer cells to DNA-damaging agents (DDAs). We recently resolved the underlying mechanism, finding that it involves SLFN11-mediated cleavage of select tRNAs predominantly employed in the translation of the ATR and ATM Ser/Thr kinases, thereby establishing SLFN11 as a novel tRNA endonuclease. Even though SLFN11 is thus involved in two of the most prominent diseases of our time, cancer and HIV infection, its regulation remained thus far unresolved. Using MS and bioinformatics-based approaches combined with site-directed mutagenesis, we show here that SLFN11 is phosphorylated at three different sites, which requires dephosphorylation for SLFN11 to become fully functionally active. Furthermore, we identified protein phosphatase 1 catalytic subunit γ (PPP1CC) as the upstream enzyme whose activity is required for SLFN11 to cleave tRNAs and thereby act as a selective translational inhibitor. In summary, our work has identified both the mechanism of SLFN11 activation and PPP1CC as the enzyme responsible for its activation. Our findings open up future studies of the PPP1CC subunit(s) involved in SLFN11 activation and the putative kinase(s) that inactivates SLFN11.
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.