Merck
HIF-transcribed p53 chaperones HIF-1α.
Nucleic acids research (2019-09-21)
Esha Madan, Taylor M Parker, Christopher J Pelham, Antonio M Palma, Maria L Peixoto, Masaki Nagane, Aliya Chandaria, Ana R Tomás, Rita Canas-Marques, Vanessa Henriques, Antonio Galzerano, Joaquim Cabral-Teixeira, Karuppaiyah Selvendiran, Periannan Kuppusamy, Carlos Carvalho, Antonio Beltran, Eduardo Moreno, Uttam K Pati, Rajan Gogna
ABSTRACT
Chronic hypoxia is associated with a variety of physiological conditions such as rheumatoid arthritis, ischemia/reperfusion injury, stroke, diabetic vasculopathy, epilepsy and cancer. At the molecular level, hypoxia manifests its effects via activation of HIF-dependent transcription. On the other hand, an important transcription factor p53, which controls a myriad of biological functions, is rendered transcriptionally inactive under hypoxic conditions. p53 and HIF-1α are known to share a mysterious relationship and play an ambiguous role in the regulation of hypoxia-induced cellular changes. Here we demonstrate a novel pathway where HIF-1α transcriptionally upregulates both WT and MT p53 by binding to five response elements in p53 promoter. In hypoxic cells, this HIF-1α-induced p53 is transcriptionally inefficient but is abundantly available for protein-protein interactions. Further, both WT and MT p53 proteins bind and chaperone HIF-1α to stabilize its binding at its downstream DNA response elements. This p53-induced chaperoning of HIF-1α increases synthesis of HIF-regulated genes and thus the efficiency of hypoxia-induced molecular changes. This basic biology finding has important implications not only in the design of anti-cancer strategies but also for other physiological conditions where hypoxia results in disease manifestation.
Sigma-Aldrich
MISSION® shRNA, Bacterial Stock
Sigma-Aldrich
Anti-p53 (Ab-3) (Mutant) Mouse mAb (PAb240), lyophilized, clone PAb240, Calbiochem®
Sigma-Aldrich
Anti-p53 (Ab-5) (Wild type) Mouse mAb (PAb1620), liquid, clone PAb1620, Calbiochem®