Merck
  • Comparing effects of rest with or without a NK1RA on fibrosis and sensorimotor declines induced by a voluntary moderate demand task.
Comparing effects of rest with or without a NK1RA on fibrosis and sensorimotor declines induced by a voluntary moderate demand task.
Journal of musculoskeletal & neuronal interactions (2019-12-04)
Mary F Barbe, Amanda R White, Brendan A Hilliard, Danielle M Salvadeo, Mamta Amin, Michele Y Harris, Geneva E Cruz, Lucas Hobson, Steven N Popoff
ABSTRACT
Fibrosis is one contributing factor in motor dysfunction and discomfort in patients with overuse musculoskeletal disorders. We pharmacologically targeted the primary receptor for Substance P, neurokinin-1, using a specific antagonist (NK1RA) in a rat model of overuse with the goal of improving tissue fibrosis and discomfort. Female rats performed a low repetition, high force (LRHF) grasping task for 12 weeks, or performed the task for 12 weeks before being placed on a four week rest break, with or without simultaneous NK1RA treatment. Results were compared to control rats (untreated, or treated 4 weeks with NK1RA or vehicle). Rest improved LRHF-induced declines in grip strength, although rest plus NK1RA treatment (Rest/NK1RA) rescued it. Both treatments improved LRHF-induced increases in muscle TGFβ1 and collagen type 1 levels, forepaw mechanical hypersensitivity (Rest/NK1RA more effectively), macrophage influx into median nerves, and enhanced collagen deposition in forepaw dermis. Only Rest/NK1RA reduced muscle hypercellularity. However, LRHF+4wk Rest /NK1RA rats showed hyposensitivity to noxious hot temperatures. While the NK1RA induced hot temperature hyposensitivity should be taken into consideration if this or related drug were used long-term, the NK1RA more effectively reduced muscle hypercellularity and improved grip strength and forepaw mechanical hypersensitivity.
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Millipore
MILLIPLEX® Rat Vascular Injury Magnetic Bead Panel 1 - Toxicity Multiplex Assay, The analytes available for this multiplex kit are: Caveolin-1, CINC-1/GRO/KC, CTGF (Connective Tissue Growth Factor), IL-6, MCP-1, PAI-1 (total), TIMP-1, TNFα, VEGF.