Merck
  • The E3 Ubiquitin Ligase Mind Bomb 1 Controls Adenovirus Genome Release at the Nuclear Pore Complex.
The E3 Ubiquitin Ligase Mind Bomb 1 Controls Adenovirus Genome Release at the Nuclear Pore Complex.
Cell reports (2019-12-19)
Michael Bauer, Justin W Flatt, Daria Seiler, Bettina Cardel, Mario Emmenlauer, Karin Boucke, Maarit Suomalainen, Silvio Hemmi, Urs F Greber
ABSTRACT
Adenoviruses (AdVs) cause respiratory, ocular, and gastrointestinal tract infection and inflammation in immunocompetent people and life-threatening disease upon immunosuppression. AdV vectors are widely used in gene therapy and vaccination. Incoming particles attach to nuclear pore complexes (NPCs) of post-mitotic cells, then rupture and deliver viral DNA (vDNA) to the nucleus or misdeliver to the cytosol. Our genome-wide RNAi screen in AdV-infected cells identified the RING-type E3 ubiquitin ligase Mind bomb 1 (Mib1) as a proviral host factor for AdV infection. Mib1 is implicated in Notch-Delta signaling, ciliary biogenesis, and RNA innate immunity. Mib1 depletion arrested incoming AdVs at NPCs. Induced expression of full-length but not ligase-defective Mib1 in knockout cells triggered vDNA uncoating from NPC-tethered virions, nuclear import, misdelivery of vDNA, and vDNA expression. Mib1 is an essential host factor for AdV uncoating in human cells, and it provides a new concept for licensing virion DNA delivery through the NPC.
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Aminoguanidine hydrochloride, ≥98%
Sigma-Aldrich
Anti-Adenovirus Antibody, clone 20/11, clone 20/11, Chemicon®, from mouse
Sigma-Aldrich
Copper(II) sulfate, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Tris(3-hydroxypropyltriazolylmethyl)amine, 95%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-E1A (Ad2/Ad5) Antibody, clone M73, clone M73, Upstate®, from mouse