Merck
  • The Neuropeptide Galanin Is Required for Homeostatic Rebound Sleep following Increased Neuronal Activity.
The Neuropeptide Galanin Is Required for Homeostatic Rebound Sleep following Increased Neuronal Activity.
Neuron (2019-09-21)
Sabine Reichert, Oriol Pavón Arocas, Jason Rihel
ABSTRACT
Sleep pressure increases during wake and dissipates during sleep, but the molecules and neurons that measure homeostatic sleep pressure remain poorly understood. We present a pharmacological assay in larval zebrafish that generates short-term increases in wakefulness followed by sustained rebound sleep after washout. The intensity of global neuronal activity during drug-induced wakefulness predicted the amount of subsequent rebound sleep. Whole-brain mapping with the neuronal activity marker phosphorylated extracellular signal-regulated kinase (pERK) identified preoptic Galanin (Galn)-expressing neurons as selectively active during rebound sleep, and the relative induction of galn transcripts was predictive of total rebound sleep time. Galn is required for sleep homeostasis, as galn mutants almost completely lacked rebound sleep following both pharmacologically induced neuronal activity and physical sleep deprivation. These results suggest that Galn plays a key role in responding to sleep pressure signals derived from neuronal activity and functions as an output arm of the vertebrate sleep homeostat.
Sigma-Aldrich
4-Aminopyridine, ≥99%
Sigma-Aldrich
Anti-Galanin Antibody, serum, Chemicon®
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Roche
Anti-Digoxigenin-POD, Fab fragments, from sheep