Merck
pH Biosensing by PI4P Regulates Cargo Sorting at the TGN.
Developmental cell (2020-01-14)
John J H Shin, Peter Liu, Leslie J Chan, Azmat Ullah, Jingxi Pan, Christoph H Borchers, John E Burke, Christopher Stefan, Gertien J Smits, Christopher J R Loewen
ABSTRACT
Phosphoinositides, diacylglycerolpyrophosphate, ceramide-1-phosphate, and phosphatidic acid belong to a unique class of membrane signaling lipids that contain phosphomonoesters in their headgroups having pKa values in the physiological range. The phosphomonoester headgroup of phosphatidic acid enables this lipid to act as a pH biosensor as changes in its protonation state with intracellular pH regulate binding to effector proteins. Here, we demonstrate that binding of pleckstrin homology (PH) domains to phosphatidylinositol 4-phosphate (PI4P) in the yeast trans-Golgi network (TGN) is dependent on intracellular pH, indicating PI4P is a pH biosensor. pH biosensing by TGN PI4P in response to nutrient availability governs protein sorting at the TGN, likely by regulating sterol transfer to the TGN by Osh1, a member of the conserved oxysterol-binding protein (OSBP) family of lipid transfer proteins. Thus, pH biosensing by TGN PI4P allows for direct metabolic regulation of protein trafficking and cell growth.
Sigma-Aldrich
Phenylarsine oxide, ≥97%, powder
Sigma-Aldrich
Guanosine 5′-diphosphate sodium salt, Type I, ≥96% (HPLC)
Avanti
C16-18:1 PC, Avanti Research - A Croda Brand 878112P, powder
Sigma-Aldrich
L-Aspartic acid β-hydroxamate, serine racemase inhibitor
Avanti
C16-18:1 PC, Avanti Research - A Croda Brand 878112C