Merck
  • Transcription factor Ctip2 controls epidermal lipid metabolism and regulates expression of genes involved in sphingolipid biosynthesis during skin development.
Transcription factor Ctip2 controls epidermal lipid metabolism and regulates expression of genes involved in sphingolipid biosynthesis during skin development.
The Journal of investigative dermatology (2012-10-26)
Zhixing Wang, Jay S Kirkwood, Alan W Taylor, Jan F Stevens, Mark Leid, Gitali Ganguli-Indra, Arup K Indra
ABSTRACT
The stratum corneum is composed of protein-enriched corneocytes embedded in an intercellular matrix of nonpolar lipids organized as lamellar layers and giving rise to epidermal permeability barrier (EPB). EPB defects have an important role in the pathophysiology of skin diseases such as eczema. The transcriptional control of skin lipid metabolism is poorly understood. We have discovered that mice lacking transcription factor COUP-TF-interacting protein 2 (Ctip2) exhibit EPB defects including altered keratinocyte terminal differentiation, delayed skin barrier development, and interrupted neutral lipid distribution in the epidermis. Here we adapted a targeted lipidomic approach using mass spectrometry and have determined that Ctip2(-/-) mice (germline deletion of the Ctip2 gene) display altered composition of major epidermal lipids, such as ceramides and sphingomyelins, compared with wild-type mice at different stages of skin development. Interestingly, expressions of several genes involved in skin sphingolipid biosynthesis and metabolism were altered in mutant skin. Ctip2 was found to be recruited to the promoter region of a subset of those genes, suggesting their possible direct regulation by Ctip2. Our results confirm an important role of Ctip2 in regulating skin lipid metabolism and indicate that profiling of epidermal sphingolipid could be useful for designing effective strategies to improve barrier dysfunctions.
Product Description
Avanti
Sphingosine (d18:1), Avanti Research - A Croda Brand
Avanti
C18:1 Ceramide (d18:1/18:1(9Z)), Avanti Research - A Croda Brand 860519P, powder
Avanti
18:0 SM (d18:1/18:0), Avanti Research - A Croda Brand 860586P, powder
Avanti
24:0 SM, Avanti Research - A Croda Brand 860592P, powder
Avanti
24:1 SM, Avanti Research - A Croda Brand, powder
Avanti
C16 Ceramide (d18:1/16:0), Avanti Research - A Croda Brand
Avanti
18:1 SM (d18:1/18:1(9Z)), Avanti Research - A Croda Brand 860587C
Avanti
C24:1 Ceramide (d18:1/24:1(15Z)), Avanti Research - A Croda Brand
Avanti
Sphinganine (d18:0), Avanti Research - A Croda Brand
Avanti
C10 Ceramide (d18:1/10:0), Avanti Research - A Croda Brand 860510P, powder
Avanti
18:1 SM (d18:1/18:1(9Z)), Avanti Research - A Croda Brand 860587P, powder