Merck
  • Acyl-CoA-Binding Protein Is a Lipogenic Factor that Triggers Food Intake and Obesity.
Acyl-CoA-Binding Protein Is a Lipogenic Factor that Triggers Food Intake and Obesity.
Cell metabolism (2019-08-20)
José M Bravo-San Pedro, Valentina Sica, Isabelle Martins, Jonathan Pol, Friedemann Loos, Maria Chiara Maiuri, Sylvère Durand, Noélie Bossut, Fanny Aprahamian, Gerasimos Anagnostopoulos, Mireia Niso-Santano, Fernando Aranda, Ignacio Ramírez-Pardo, Justine Lallement, Jessica Denom, Erwan Boedec, Philip Gorwood, Nicolas Ramoz, Karine Clément, Veronique Pelloux, Alili Rohia, François Pattou, Violeta Raverdy, Robert Caiazzo, Raphaël G P Denis, Patricia Boya, Lorenzo Galluzzi, Frank Madeo, Stéphanie Migrenne-Li, Céline Cruciani-Guglielmacci, Nektarios Tavernarakis, Carlos López-Otín, Christophe Magnan, Guido Kroemer
ABSTRACT
Autophagy facilitates the adaptation to nutritional stress. Here, we show that short-term starvation of cultured cells or mice caused the autophagy-dependent cellular release of acyl-CoA-binding protein (ACBP, also known as diazepam-binding inhibitor, DBI) and consequent ACBP-mediated feedback inhibition of autophagy. Importantly, ACBP levels were elevated in obese patients and reduced in anorexia nervosa. In mice, systemic injection of ACBP protein inhibited autophagy, induced lipogenesis, reduced glycemia, and stimulated appetite as well as weight gain. We designed three approaches to neutralize ACBP, namely, inducible whole-body knockout, systemic administration of neutralizing antibodies, and induction of antiACBP autoantibodies in mice. ACBP neutralization enhanced autophagy, stimulated fatty acid oxidation, inhibited appetite, reduced weight gain in the context of a high-fat diet or leptin deficiency, and accelerated weight loss in response to dietary changes. In conclusion, neutralization of ACBP might constitute a strategy for treating obesity and its co-morbidities.
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Ponceau S solution, BioReagent, suitable (for use in cellulose acetate electrophoresis), 0.1 % (w/v) in 5% acetic acid
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Oil Red O solution, 0.5% in isopropanol
Sigma-Aldrich
Brefeldin A, BioXtra, ≥99% (HPLC and TLC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
N-[Tris(hydroxymethyl)methyl]acrylamide, contains ≤7% KCl, 93%
Sigma-Aldrich
Perifosine, ≥98% (HPLC)