Merck
  • Exosomes derived from microRNA-30b-3p-overexpressing mesenchymal stem cells protect against lipopolysaccharide-induced acute lung injury by inhibiting SAA3.
Exosomes derived from microRNA-30b-3p-overexpressing mesenchymal stem cells protect against lipopolysaccharide-induced acute lung injury by inhibiting SAA3.
Experimental cell research (2019-06-07)
Xiaomeng Yi, Xuxia Wei, Haijin Lv, Yuling An, Lijuan Li, Pinglan Lu, Yang Yang, Qi Zhang, Huimin Yi, Guihua Chen
ABSTRACT
Mesenchymal stem cells (MSCs) have been widely documented for their potential role in the treatment of various clinical disorders, including acute lung injury (ALI). ALI represents a clinical syndrome associated with histopathological diffuse alveolar damage. Recent evidence has demonstrated that exosomes derived from MSCs may serve as a reservoir of anti-apoptotic microRNAs (miRs) conferring protection from certain diseases. Hence, the current study was performed with the aim of investigating whether MSCs-exosomal miR-30b-3p could confer protection against ALI. A bioinformatic analysis and a dual luciferase assay were initially performed to verify that SAA3 was highly-expressed in ALI which was confirmed to be a target gene of miR-30b-3p. Next, the lipopolysaccharide (LPS)-treated type II alveolar epithelial cells (AECs) (MLE-12) were transfected with mimics or inhibitors of miR-30b-3p, or sh-SAA3. It was revealed that LPS induced AEC apoptosis, which could be inhibited by overexpressing miR-30b-3p by down-regulating the expression of SAA3. After co-culture of PKH26-labeled exosomes with MLE-12 cells, we found that the number of PKH26-labeled exosomes endocytosed by MLE-12 cells gradually increased. Furthermore, the LPS-treated MLE-12 cells co-cultured with MSC-exosomes overexpressing miR-30b-3p exhibited increased miR-30b-3p, decreased SAA3 level, as well as increased cell proliferation, accompanied by diminished cell apoptosis in LPS-treated MLE-12 cells. Finally, the protective effect of MSCs-exosomal miR-30b-3p on the AECs in vivo was investigated in an ALI mouse model with tail vein injection of MSC-exosomes with elevated miR-30b-3p, showing that overexpression of miR-30b-3p in MSC-exosomes conferred protective effects against ALI. Taken together, these findings highlighted the potential of MSC-exosomes overexpressing miR-30b-3p in preventing ALI. The exosomes derived from MSCs hold potential as future therapeutic strategies in the treatment of ALI.