Merck
  • Dynamic Incorporation of Histone H3 Variants into Chromatin Is Essential for Acquisition of Aggressive Traits and Metastatic Colonization.
Dynamic Incorporation of Histone H3 Variants into Chromatin Is Essential for Acquisition of Aggressive Traits and Metastatic Colonization.
Cancer cell (2019-10-01)
Ana P Gomes, Didem Ilter, Vivien Low, Adam Rosenzweig, Zih-Jie Shen, Tanya Schild, Martin A Rivas, Ekrem E Er, Dylan R McNally, Anders P Mutvei, Julie Han, Yi-Hung Ou, Paola Cavaliere, Edouard Mullarky, Michal Nagiec, Sejeong Shin, Sang-Oh Yoon, Noah Dephoure, Joan Massagué, Ari M Melnick, Lewis C Cantley, Jessica K Tyler, John Blenis
ABSTRACT
Metastasis is the leading cause of cancer mortality. Chromatin remodeling provides the foundation for the cellular reprogramming necessary to drive metastasis. However, little is known about the nature of this remodeling and its regulation. Here, we show that metastasis-inducing pathways regulate histone chaperones to reduce canonical histone incorporation into chromatin, triggering deposition of H3.3 variant at the promoters of poor-prognosis genes and metastasis-inducing transcription factors. This specific incorporation of H3.3 into chromatin is both necessary and sufficient for the induction of aggressive traits that allow for metastasis formation. Together, our data clearly show incorporation of histone variant H3.3 into chromatin as a major regulator of cell fate during tumorigenesis, and histone chaperones as valuable therapeutic targets for invasive carcinomas.
Sigma-Aldrich
Carboplatin
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Anti-Histone H3.1/H3.2 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
ChIPAb+ Histone H3.3 - ChIP Validated Antibody and Primer Set, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Sodium cyanoborodeuteride, 97 atom % D, ≥96% (CP)
Sigma-Aldrich
Tetrapropylammonium perruthenate, 97%
Sigma-Aldrich
Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Hexadimethrine bromide, ≥94% (titration)
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, ≥98% (HPLC), powder
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)
Sigma-Aldrich
Anti-Histone H3.3 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
MISSION® esiRNA, targeting human CHAF1B
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Donkey Anti-Goat IgG Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from donkey