Merck
  • Structural Organization and Dynamics of Homodimeric Cytohesin Family Arf GTPase Exchange Factors in Solution and on Membranes.
Structural Organization and Dynamics of Homodimeric Cytohesin Family Arf GTPase Exchange Factors in Solution and on Membranes.
Structure (London, England : 1993) (2019-10-12)
Sanchaita Das, Andrew W Malaby, Agata Nawrotek, Wenhua Zhang, Mahel Zeghouf, Sarah Maslen, Mark Skehel, Srinivas Chakravarthy, Thomas C Irving, Osman Bilsel, Jacqueline Cherfils, David G Lambright
ABSTRACT
Membrane dynamic processes require Arf GTPase activation by guanine nucleotide exchange factors (GEFs) with a Sec7 domain. Cytohesin family Arf GEFs function in signaling and cell migration through Arf GTPase activation on the plasma membrane and endosomes. In this study, the structural organization of two cytohesins (Grp1 and ARNO) was investigated in solution by size exclusion-small angle X-ray scattering and negative stain-electron microscopy and on membranes by dynamic light scattering, hydrogen-deuterium exchange-mass spectrometry and guanosine diphosphate (GDP)/guanosine triphosphate (GTP) exchange assays. The results suggest that cytohesins form elongated dimers with a central coiled coil and membrane-binding pleckstrin-homology (PH) domains at opposite ends. The dimers display significant conformational heterogeneity, with a preference for compact to intermediate conformations. Phosphoinositide-dependent membrane recruitment is mediated by one PH domain at a time and alters the conformational dynamics to prime allosteric activation by Arf-GTP. A structural model for membrane targeting and allosteric activation of full-length cytohesin dimers is discussed.
Product Description
Avanti
Brain PC, Avanti Research - A Croda Brand 840053C
Pricing and availability is not currently available.
Avanti
Brain PC, Avanti Research - A Croda Brand 840053P, powder
Pricing and availability is not currently available.