Merck
  • Central nicotine induces browning through hypothalamic κ opioid receptor.
Central nicotine induces browning through hypothalamic κ opioid receptor.
Nature communications (2019-09-08)
Patricia Seoane-Collazo, Laura Liñares-Pose, Eva Rial-Pensado, Amparo Romero-Picó, José María Moreno-Navarrete, Noelia Martínez-Sánchez, Pablo Garrido-Gil, Ramón Iglesias-Rey, Donald A Morgan, Naoki Tomasini, Samuel Andrew Malone, Ana Senra, Cintia Folgueira, Gema Medina-Gomez, Tomás Sobrino, José L Labandeira-García, Rubén Nogueiras, Ana I Domingos, José-Manuel Fernández-Real, Kamal Rahmouni, Carlos Diéguez, Miguel López
ABSTRACT
Increased body weight is a major factor that interferes with smoking cessation. Nicotine, the main bioactive compound in tobacco, has been demonstrated to have an impact on energy balance, since it affects both feeding and energy expenditure at the central level. Among the central actions of nicotine on body weight, much attention has been focused on its effect on brown adipose tissue (BAT) thermogenesis, though its effect on browning of white adipose tissue (WAT) is unclear. Here, we show that nicotine induces the browning of WAT through a central mechanism and that this effect is dependent on the κ opioid receptor (KOR), specifically in the lateral hypothalamic area (LHA). Consistent with these findings, smokers show higher levels of uncoupling protein 1 (UCP1) expression in WAT, which correlates with smoking status. These data demonstrate that central nicotine-induced modulation of WAT browning may be a target against human obesity.
Sigma-Aldrich
Anti-AMPK α1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-AMPK α2 Antibody, Upstate®, from rabbit