Merck
  • 12-Lipoxygenase Regulates Cold Adaptation and Glucose Metabolism by Producing the Omega-3 Lipid 12-HEPE from Brown Fat.
12-Lipoxygenase Regulates Cold Adaptation and Glucose Metabolism by Producing the Omega-3 Lipid 12-HEPE from Brown Fat.
Cell metabolism (2019-07-30)
Luiz Osório Leiria, Chih-Hao Wang, Matthew D Lynes, Kunyan Yang, Farnaz Shamsi, Mari Sato, Satoru Sugimoto, Emily Y Chen, Valerie Bussberg, Niven R Narain, Brian E Sansbury, Justin Darcy, Tian Lian Huang, Sean D Kodani, Masaji Sakaguchi, Andréa L Rocha, Tim J Schulz, Alexander Bartelt, Gökhan S Hotamisligil, Michael F Hirshman, Klaus van Leyen, Laurie J Goodyear, Matthias Blüher, Aaron M Cypess, Michael A Kiebish, Matthew Spite, Yu-Hua Tseng
ABSTRACT
Distinct oxygenases and their oxylipin products have been shown to participate in thermogenesis by mediating physiological adaptations required to sustain body temperature. Since the role of the lipoxygenase (LOX) family in cold adaptation remains elusive, we aimed to investigate whether, and how, LOX activity is required for cold adaptation and to identify LOX-derived lipid mediators that could serve as putative cold mimetics with therapeutic potential to combat diabetes. By utilizing mass-spectrometry-based lipidomics in mice and humans, we demonstrated that cold and β3-adrenergic stimulation could promote the biosynthesis and release of 12-LOX metabolites from brown adipose tissue (BAT). Moreover, 12-LOX ablation in mouse brown adipocytes impaired glucose uptake and metabolism, resulting in blunted adaptation to the cold in vivo. The cold-induced 12-LOX product 12-HEPE was found to be a batokine that improves glucose metabolism by promoting glucose uptake into adipocytes and skeletal muscle through activation of an insulin-like intracellular signaling pathway.
Sigma-Aldrich
Barium hydroxide solution, 0.3 N
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Insulin from bovine pancreas, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(±)-Norepinephrine (+)-bitartrate salt
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
NCTT-956, ≥97% (NMR)
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Oligomycin, A mixture of A, B, and C isomers.
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Zinc sulfate solution, 0.3 N
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%