Merck
  • Enhanced sensitivity to hydrogen peroxide-induced apoptosis in Evi1 transformed Rat1 fibroblasts due to repression of carbonic anhydrase III.
Enhanced sensitivity to hydrogen peroxide-induced apoptosis in Evi1 transformed Rat1 fibroblasts due to repression of carbonic anhydrase III.
The FEBS journal (2009-12-18)
P Roy, E Reavey, M Rayne, S Roy, M Abed El Baky, Y Ishii, C Bartholomew
ABSTRACT
EVI1 is a nuclear zinc finger protein essential to normal development, which participates in acute myeloid leukaemia progression and transforms Rat1 fibroblasts. In this study we show that enforced expression of Evi1 in Rat1 fibroblasts protects from paclitaxel-induced apoptosis, consistent with previously published studies. Surprisingly, however, these cells show increased sensitivity to hydrogen peroxide (H(2)O(2))-induced apoptosis, demonstrated by elevated caspase 3 catalytic activity. This effect is caused by a reduction in carbonic anhydrase III (caIII) production. caIII transcripts are repressed by 92-97% by Evi1 expression, accompanied by a similar reduction in caIII protein. Reporter assays with the rat caIII gene promoter show repressed activity, demonstrating that Evi1 either directly or indirectly modulates transcription of this gene in Rat1 cells. Targeted knockdown of caIII alone, with Dicer-substrate short inhibitory RNAs, also increases the sensitivity of Rat1 fibroblasts to H(2)O(2), which occurs in the absence of any other changes mediated by Evi1 expression. Enforced expression of caIII in Evi1-expressing Rat1 cells reverts the phenotype, restoring H(2)O(2) resistance. Together these data show that Evi1 represses transcription of caIII gene expression, leading to increased sensitivity to H(2)O(2)-induced apoptosis in Rat1 cells and might suggest the basis for the development of a novel therapeutic strategy for the treatment of leukaemias and solid tumours where EVI1 is overexpressed.
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
DCFHDA, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Pricing and availability is not currently available.