Merck
  • Membrane-cytoskeletal crosstalk mediated by myosin-I regulates adhesion turnover during phagocytosis.
Membrane-cytoskeletal crosstalk mediated by myosin-I regulates adhesion turnover during phagocytosis.
Nature communications (2019-03-21)
Sarah R Barger, Nicholas S Reilly, Maria S Shutova, Qingsen Li, Paolo Maiuri, John M Heddleston, Mark S Mooseker, Richard A Flavell, Tatyana Svitkina, Patrick W Oakes, Mira Krendel, Nils C Gauthier
ABSTRACT
Phagocytosis of invading pathogens or cellular debris requires a dramatic change in cell shape driven by actin polymerization. For antibody-covered targets, phagocytosis is thought to proceed through the sequential engagement of Fc-receptors on the phagocyte with antibodies on the target surface, leading to the extension and closure of the phagocytic cup around the target. We find that two actin-dependent molecular motors, class 1 myosins myosin 1e and myosin 1f, are specifically localized to Fc-receptor adhesions and required for efficient phagocytosis of antibody-opsonized targets. Using primary macrophages lacking both myosin 1e and myosin 1f, we find that without the actin-membrane linkage mediated by these myosins, the organization of individual adhesions is compromised, leading to excessive actin polymerization, slower adhesion turnover, and deficient phagocytic internalization. This work identifies a role for class 1 myosins in coordinated adhesion turnover during phagocytosis and supports a mechanism involving membrane-cytoskeletal crosstalk for phagocytic cup closure.
Sigma-Aldrich
Anti-F-actin-capping protein subunit beta Antibody, from rabbit
Sigma-Aldrich
Anti-Arp2/3 complex Antibody, clone 13C9, clone 13C9, from mouse