Merck
  • Potent and PPARα-independent anti-proliferative action of the hypolipidemic drug fenofibrate in VEGF-dependent angiosarcomas in vitro.
Potent and PPARα-independent anti-proliferative action of the hypolipidemic drug fenofibrate in VEGF-dependent angiosarcomas in vitro.
Scientific reports (2019-04-21)
Yasser Majeed, Rohit Upadhyay, Sara Alhousseiny, Tarek Taha, Adham Musthak, Yanal Shaheen, Mohtashim Jameel, Chris R Triggle, Hong Ding
ABSTRACT
Angiosarcomas are highly aggressive tumors of endothelial origin, which carry a poor prognosis. Fenofibrate is a hypolipidemic drug, which acts by activating the transcription factor PPARα. It has also been widely reported to have 'anti-cancer' activity. The current study investigated its effect in a murine VEGF-dependent angiosarcoma cell-line, MS1 VEGF. The study utilised assays to monitor cell proliferation and viability, apoptosis, cell cycle progression, mitochondrial membrane potential, changes in protein expression, and changes in miRNA expression using microarrays. Fenofibrate showed potent anti-proliferative action in MS1 VEGF angiosarcoma cells, without inducing apoptosis. It enriched cells in G2/M cell cycle phase and hyperpolarised mitochondria. Other PPARα activators failed to mimic fenofibrate action. Inhibitors of PPARα and NFκB failed to reverse the inhibitory effect of fenofibrate and their combination with fenofibrate was cytotoxic. Fenofibrate downregulated the expression of key VEGF-effector proteins, including Akt, ERK, Bcl-2 and survivin, and a chemical inhibitor screen discovered relevance of these proteins to cell proliferation. A miRNA microarray revealed that fenofibrate differentially regulated cellular miRNAs with known roles in cancer and angiogenesis. The data raise the possibility that fenofibrate could be useful in angiosarcoma therapy, especially considering its well-established clinical safety and tolerability profile.
Sigma-Aldrich
SU-5402, ≥98% (HPLC)
Sigma-Aldrich
PD 98,059, solid
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
Ammonium pyrrolidinedithiocarbamate, ~99%
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)