Merck
  • Metabolic and Innate Immune Cues Merge into a Specific Inflammatory Response via the UPR.
Metabolic and Innate Immune Cues Merge into a Specific Inflammatory Response via the UPR.
Cell (2019-04-30)
Denis A Mogilenko, Joel T Haas, Laurent L'homme, Sébastien Fleury, Sandrine Quemener, Matthieu Levavasseur, Coralie Becquart, Julien Wartelle, Alexandra Bogomolova, Laurent Pineau, Olivier Molendi-Coste, Steve Lancel, Hélène Dehondt, Celine Gheeraert, Aurelie Melchior, Cédric Dewas, Artemii Nikitin, Samuel Pic, Nabil Rabhi, Jean-Sébastien Annicotte, Seiichi Oyadomari, Talia Velasco-Hernandez, Jörg Cammenga, Marc Foretz, Benoit Viollet, Milica Vukovic, Arnaud Villacreces, Kamil Kranc, Peter Carmeliet, Guillemette Marot, Alexis Boulter, Simon Tavernier, Luciana Berod, Maria P Longhi, Christophe Paget, Sophie Janssens, Delphine Staumont-Sallé, Ezra Aksoy, Bart Staels, David Dombrowicz
ABSTRACT
Innate immune responses are intricately linked with intracellular metabolism of myeloid cells. Toll-like receptor (TLR) stimulation shifts intracellular metabolism toward glycolysis, while anti-inflammatory signals depend on enhanced mitochondrial respiration. How exogenous metabolic signals affect the immune response is unknown. We demonstrate that TLR-dependent responses of dendritic cells (DCs) are exacerbated by a high-fatty-acid (FA) metabolic environment. FAs suppress the TLR-induced hexokinase activity and perturb tricarboxylic acid cycle metabolism. These metabolic changes enhance mitochondrial reactive oxygen species (mtROS) production and, in turn, the unfolded protein response (UPR), leading to a distinct transcriptomic signature with IL-23 as hallmark. Interestingly, chemical or genetic suppression of glycolysis was sufficient to induce this specific immune response. Conversely, reducing mtROS production or DC-specific deficiency in XBP1 attenuated IL-23 expression and skin inflammation in an IL-23-dependent model of psoriasis. Thus, fine-tuning of innate immunity depends on optimization of metabolic demands and minimization of mtROS-induced UPR.
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
MitoTEMPO, ≥98% (HPLC)
Sigma-Aldrich
(+)-Etomoxir sodium salt hydrate, ≥98% (HPLC), powder