Merck
  • Enhanced Antioxidant Activity for Apple Juice Fermented with Lactobacillus plantarum ATCC14917.
Enhanced Antioxidant Activity for Apple Juice Fermented with Lactobacillus plantarum ATCC14917.
Molecules (Basel, Switzerland) (2018-12-28)
Zhongxi Li, Jing Teng, Yilu Lyu, Xiaoqian Hu, Yueliang Zhao, Mingfu Wang
ABSTRACT
The present study examined the effect of Lactobacillus plantarum ATCC14917 fermentation on the chemical composition and antioxidant activity of apple juice. Apple juice was fermented and examined of its antioxidant activity using chemical models and cellular antioxidant assay. Furthermore, the chemical composition of fermented apple juice was characterized by LC-MS/MS. Lactobacillus plantarum ATCC14917 fermentation showed an increase in DPPH and ABTS radical scavenging activity as well as cellular antioxidant activity of apple juice. However, fermentation decreased the total phenolic and flavonoid content. Subsequent LC-MS/MS analysis of the phenolic profile indicated that the content of 5-O-caffeoylquinic acid (5-CQA), quercetin, and phloretin with strong antioxidant activity was increased significantly after fermentation. The modified phenolic composition may contribute to the increased antioxidant activity of fermented apple juice. Our findings showed that Lactobacillus plantarum ATCC14917 fermentation may be an efficient way to enhance the bioavailability of phenolic compounds and to protect cells from oxidative stress.
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Ellagic acid, ≥95% (HPLC), powder, from tree bark