Merck
  • Rapid pathology of lumpectomy margins with open-top light-sheet (OTLS) microscopy.
Rapid pathology of lumpectomy margins with open-top light-sheet (OTLS) microscopy.
Biomedical optics express (2019-03-21)
Ye Chen, Weisi Xie, Adam K Glaser, Nicholas P Reder, Chenyi Mao, Suzanne M Dintzis, Joshua C Vaughan, Jonathan T C Liu
ABSTRACT
Open-top light-sheet microscopy is a technique that can potentially enable rapid ex vivo inspection of large tissue surfaces and volumes. Here, we have optimized an open-top light-sheet (OTLS) microscope and image-processing workflow for the comprehensive examination of surgical margin surfaces, and have also developed a novel fluorescent analog of H&E staining that is robust for staining fresh unfixed tissues. Our tissue-staining method can be achieved within 2.5 minutes followed by OTLS microscopy of lumpectomy surfaces at a rate of up to 1.5 cm2/minute. An image atlas is presented to show that OTLS image quality surpasses that of intraoperative frozen sectioning and can approximate that of gold-standard H&E histology of formalin-fixed paraffin-embedded (FFPE) tissues. Qualitative evidence indicates that these intraoperative methods do not interfere with downstream post-operative H&E histology and immunohistochemistry. These results should facilitate the translation of OTLS microscopy for intraoperative guidance of lumpectomy and other surgical oncology procedures.