Merck
  • ANP32 Proteins Are Essential for Influenza Virus Replication in Human Cells.
ANP32 Proteins Are Essential for Influenza Virus Replication in Human Cells.
Journal of virology (2019-06-21)
Ecco Staller, Carol M Sheppard, Peter J Neasham, Bhakti Mistry, Thomas P Peacock, Daniel H Goldhill, Jason S Long, Wendy S Barclay
ABSTRACT
ANP32 proteins have been implicated in supporting influenza virus replication, but most of the work to date has focused on the ability of avian Anp32 proteins to overcome restriction of avian influenza polymerases in human cells. Using a CRISPR approach, we show that the human acidic nuclear phosphoproteins (ANPs) ANP32A and ANP32B are functionally redundant but essential host factors for mammalian-adapted influenza A virus (IAV) and influenza B virus (IBV) replication in human cells. When both proteins are absent from human cells, influenza polymerases are unable to replicate the viral genome, and infectious virus cannot propagate. Provision of exogenous ANP32A or ANP32B recovers polymerase activity and virus growth. We demonstrate that this redundancy is absent in the murine Anp32 orthologues; murine Anp32A is incapable of recovering IAV polymerase activity, while murine Anp32B can do so. Intriguingly, IBV polymerase is able to use murine Anp32A. We show, using a domain swap and point mutations, that the leucine-rich repeat (LRR) 5 region comprises an important functional domain for mammalian ANP32 proteins. Our approach has identified a pair of essential host factors for influenza virus replication and can be harnessed to inform future interventions.IMPORTANCE Influenza virus is the etiological agent behind some of the most devastating infectious disease pandemics to date, and influenza outbreaks still pose a major threat to public health. Influenza virus polymerase, the molecule that copies the viral RNA genome, hijacks cellular proteins to support its replication. Current anti-influenza drugs are aimed against viral proteins, including the polymerase, but RNA viruses like influenza tend to become resistant to such drugs very rapidly. An alternative strategy is to design therapeutics that target the host proteins that are necessary for virus propagation. Here, we show that the human proteins ANP32A and ANP32B are essential for influenza A and B virus replication, such that in their absence cells become impervious to the virus. We map the proviral activity of ANP32 proteins to one region in particular, which could inform future intervention.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)