Merck
  • Characterization of advanced glycation end products and their receptor (RAGE) in an animal model of myocardial infarction.
Characterization of advanced glycation end products and their receptor (RAGE) in an animal model of myocardial infarction.
PloS one (2019-01-12)
Bianca de Moraes Fracasso, Juliana Oliveira Rangel, Alessandra Gonçalves Machado, Fernanda Severo Curuja, Amanda Lopes, Virgílio Olsen, Nadine Clausell, Andreia Biolo, Luis Eduardo Rohde, Michael Andrades
ABSTRACT
Circulating advanced glycation end products (AGE) and their receptor, RAGE, are increased after a myocardial infarction (MI) episode and seem to be associated with worse prognosis in patients. Despite the increasing importance of these molecules in the course of cardiac diseases, they have never been characterized in an animal model of MI. Thus, the aim of this study was to characterize AGE formation and RAGE expression in plasma and cardiac tissue during cardiac remodeling after MI in rats. Adult male Wistar rats were randomized to receive sham surgery (n = 15) or MI induction (n = 14) by left anterior descending coronary artery ligation. The MI group was stratified into two subgroups based on postoperative left ventricular ejection fraction: low (MIlowEF) and intermediate (MIintermEF). Echocardiography findings and plasma levels of AGEs, protein carbonyl, and free amines were assessed at baseline and 2, 30, and 120 days postoperatively. At the end of follow-up, the heart was harvested for AGE and RAGE evaluation. No differences were observed in AGE formation in plasma, except for a decrease in absorbance in MIlowEF at the end of follow-up. A decrease in yellowish-brown AGEs in heart homogenate was found, which was confirmed by immunodetection of N-ε-carboxymethyl-lysine. No differences could be seen in plasma RAGE levels among the groups, despite an increase in MI groups over the time. However, MI animals presented an increase of 50% in heart RAGE at the end of the follow-up. Despite the inflammatory and oxidative profile of experimental MI in rats, there was no increase in plasma AGE or RAGE levels. However, AGE levels in cardiac tissue declined. Thus, we suggest that the rat MI model should be employed with caution when studying the AGE-RAGE signaling axis or anti-AGE drugs for not reflecting previous clinical findings.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Tablets, For General Use
Pricing and availability is not currently available.