Merck
  • The Smad2/3/4 complex binds miR-139 promoter to modulate TGFβ-induced proliferation and activation of human Tenon's capsule fibroblasts through the Wnt pathway.
The Smad2/3/4 complex binds miR-139 promoter to modulate TGFβ-induced proliferation and activation of human Tenon's capsule fibroblasts through the Wnt pathway.
Journal of cellular physiology (2019-01-17)
Mi Deng, Shi-Ying Hou, Bo-Ding Tong, Jia-Yang Yin, Wei Xiong
ABSTRACT
The activation and proliferation of human Tenon's fibroblasts (HTFs) play a vital role in the fibrosis in the pathology of the scar formation after the glaucoma filtration surgery. Transforming growth factor β1 (TGFβ1)/Smads signaling has been reported to promote fibrosis. In our previous study, we revealed that TGFβ1-induced orbital fibroblast activation and proliferation through Wnt/β-catenin signaling. As microRNA (miR)-139 could target several factors in Wnt signaling to modulate fibrosis, here, the effect and mechanism of miR-139 in HTF activation and proliferation were investigated. miR-139 overexpression significantly reversed the TGFβ1-induced increase in collagen I and α-smooth muscle actin contents and proliferation in HTFs. CTNNB1 and CTNND1 were direct downstream of miR-139 and can significantly restore the suppressive effect of miR-139 on the activation and proliferation in HTFs under TGFβ1 stimulation. Smad2/3/4 complex inhibits the transcription activity of miR-139, most possibly by Smad4 binding to the miR-139 promoter. Taken together, we demonstrated a new mechanism of HTF activation and proliferation from the perspective of miRNA regulation, which may provide new strategies for improving the fibrosis after the glaucoma filtration surgery.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture