Merck
  • A mechano-signalling network linking microtubules, myosin IIA filaments and integrin-based adhesions.
A mechano-signalling network linking microtubules, myosin IIA filaments and integrin-based adhesions.
Nature materials (2019-05-23)
Nisha Bte Mohd Rafiq, Yukako Nishimura, Sergey V Plotnikov, Visalatchi Thiagarajan, Zhen Zhang, Shidong Shi, Meenubharathi Natarajan, Virgile Viasnoff, Pakorn Kanchanawong, Gareth E Jones, Alexander D Bershadsky
ABSTRACT
The interrelationship between microtubules and the actin cytoskeleton in mechanoregulation of integrin-mediated adhesions is poorly understood. Here, we show that the effects of microtubules on two major types of cell-matrix adhesion, focal adhesions and podosomes, are mediated by KANK family proteins connecting the adhesion protein talin with microtubule tips. Both total microtubule disruption and microtubule uncoupling from adhesions by manipulations with KANKs trigger a massive assembly of myosin IIA filaments, augmenting focal adhesions and disrupting podosomes. Myosin IIA filaments are indispensable effectors in the microtubule-driven regulation of integrin-mediated adhesions. Myosin IIA filament assembly depends on Rho activation by the RhoGEF GEF-H1, which is trapped by microtubules when they are connected with integrin-mediated adhesions via KANK proteins but released after their disconnection. Thus, microtubule capture by integrin-mediated adhesions modulates the GEF-H1-dependent effect of microtubules on the assembly of myosin IIA filaments. Subsequent actomyosin reorganization then remodels the focal adhesions and podosomes, closing the regulatory loop.
Sigma-Aldrich
MISSION® esiRNA, targeting human MYH10
Pricing and availability is not currently available.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human KANK2
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human KANK1
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human ARHGEF2
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human MYH9
Pricing and availability is not currently available.
Sigma-Aldrich
Triton X-100, laboratory grade
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Myosin IIA, non muscle antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Supelco
Rapamycin, VETRANAL®, analytical standard
Pricing and availability is not currently available.