Merck
VCP Machinery Mediates Autophagic Degradation of Empty Argonaute.
Cell reports (2019-08-01)
Hotaka Kobayashi, Keisuke Shoji, Kaori Kiyokawa, Lumi Negishi, Yukihide Tomari
ABSTRACT
The Argonaute subfamily of proteins (AGO) loads microRNAs (miRNAs) to form the effector complex that mediates target gene silencing. Empty AGO, but not miRNA-loaded AGO, is selectively degraded across species. We have reported that the degradation of empty AGO is part of a quality control pathway that eliminates dysfunctional AGO. However, how empty AGO is degraded remains unclear. Here we show that the empty state of Drosophila Ago1 is degraded by autophagy. Comprehensive LC-MS/MS analyses, together with manipulation of the Ago1 ubiquitination level, revealed that VCP, which mediates selective autophagy, recognizes empty Ago1 via the Ufd1-Npl4 heterodimer. Depletion of VCP-Ufd1-Npl4 machinery impairs degradation of empty Ago1 and miRNA-mediated target gene silencing. Our findings reveal a direct link between empty AGO degradation and selective autophagy that ensures efficient miRNA function.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
Anti-ATG8 Antibody, from rabbit, purified by affinity chromatography