Merck
Sonic hedgehog expression in the postnatal brain.
Biology open (2019-03-07)
Aileen Rivell, Ronald S Petralia, Ya-Xian Wang, Ellie Clawson, Keelin Moehl, Mark P Mattson, Pamela J Yao
ABSTRACT
Beyond its role in patterning the neural tube during embryogenesis, additional functions of Sonic hedgehog (Shh) in post-embryonic and mature brains have been coming into focus. However, the question of the abundance of endogenous Shh - the ligand of the signaling pathway - and its changes over time in post-embryonic and mature brains are less well understood. Here we find that while the amounts of Shh transcript and protein in rat brains are nearly undetectable at birth, they increase continuously during postnatal development and remain at readily detectable levels in young adults. This developmental age-associated increase in Shh levels is also seen in hippocampal neurons grown in culture, in which very young neurons produce minimal amounts of Shh protein but, as neurons grow and form synapses, the amounts of Shh increase significantly. Using immunolabeling with antibodies to different residues of Shh, we observed that the N-terminal fragment and the C-terminal fragment of Shh are present in hippocampal neurons, and that these two Shh forms co-exist in most compartments of the neuron. Our findings provide a better understanding of Shh expression in the brain, laying the groundwork for further comprehending the biogenesis of Shh protein in the young and mature brain and neurons.
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Sigma-Aldrich
Monoclonal Anti-MAP2 antibody produced in mouse, clone HM-2, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution