Merck
  • The synergistic effect of electroacupuncture and bone mesenchymal stem cell transplantation on repairing thin endometrial injury in rats.
The synergistic effect of electroacupuncture and bone mesenchymal stem cell transplantation on repairing thin endometrial injury in rats.
Stem cell research & therapy (2019-08-09)
Liangjun Xia, Qingyu Meng, Jin Xi, Qin Han, Jie Cheng, Jie Shen, Youbing Xia, Liyun Shi
ABSTRACT
Tissue regeneration disorder after endometrial injury is an important cause of intrauterine adhesions, amenorrhea, and infertility in women. Both bone marrow mesenchymal stem cell (BMSC) transplantation and electroacupuncture (EA) are promising therapeutic applications for endometrial injury. This study examined their combined effects on thin endometrium in rats and the possible mechanisms underlying these effects. A thin endometrial model was established in Sprague-Dawley (SD) rats by perfusing 95% ethanol into the right side of the uterus. The wounds were randomly treated with PBS (model group), BMSCs only (BMSC group), EA only (EA group), and BMSCs combined with EA (BMSC + EA group). Endometrial morphological alterations were observed by hematoxylin and eosin (H&E) staining. Changes in markers of epithelial and stromal endometrium cells, endometrial receptivity-related chemokines, and paracrine factors were detected using immunohistochemistry, western blotting, and quantitative reverse-transcription polymerase chain reaction (qRT-PCR). Finally, the functional recovery of the uterus was evaluated by determining the rate of embryo implantation. As shown by endometrial morphology, the damaged uteri in all the treatment groups recovered to some extent, with the best effects observed in the BMSC + EA group. Further studies showed that EA promoted the migration of transplanted BMSCs to damaged uteri by activating the stromal cell-derived factor-1/C-X-C chemokine receptor type 4 (SDF-1/CXCR4) axis. As compared with the other groups, upregulated expression of endometrial cytokeratin and vimentin, increased secretion of vascular endothelial growth factor (VEGF) and basic fibroblast growth factor (bFGF) in endometrial lesions, and improved embryo implantation rates on the 8th day of pregnancy were found in the BMSC + EA group. EA plays an important role in supporting BMSCs in the repair of thin endometrium, most likely by promoting the migration of BMSCs and enhancing the paracrine effect of BMSCs.