Merck
  • Lewis acid promoted highly diastereoselective Petasis Borono-Mannich reaction: efficient synthesis of optically active β,γ-unsaturated α-amino acids.
Lewis acid promoted highly diastereoselective Petasis Borono-Mannich reaction: efficient synthesis of optically active β,γ-unsaturated α-amino acids.
Organic letters (2012-04-07)
Yi Li, Ming-Hua Xu
ABSTRACT
An efficient and straightforward method for the preparation of highly enantiomerically enriched β,γ-unsaturated α-amino acid derivatives by a Lewis acid promoted diastereoselective Petasis reaction of vinylboronic acid, N-tert-butanesulfinamide, and glyoxylic acid has been developed. The synthetic utilities of the approach were demonstrated by the rapid and convenient construction of challenging cyclopenta[c]proline derivatives.
Sigma-Aldrich
Glyoxylic acid solution, 50 wt. % in H2O
Sigma-Aldrich
trans-2-Phenylvinylboronic acid, 97%