Merck
  • Cideb controls sterol-regulated ER export of SREBP/SCAP by promoting cargo loading at ER exit sites.
Cideb controls sterol-regulated ER export of SREBP/SCAP by promoting cargo loading at ER exit sites.
The EMBO journal (2019-03-13)
Lu Su, Linkang Zhou, Feng-Jung Chen, Huimin Wang, Hui Qian, Yuanyuan Sheng, Yuangang Zhu, Hua Yu, Xinqi Gong, Li'e Cai, Xuerui Yang, Li Xu, Tong-Jin Zhao, John Zhong Li, Xiao-Wei Chen, Peng Li
ABSTRACT
SREBPs are master regulators of lipid homeostasis and undergo sterol-regulated export from ER to Golgi apparatus for processing and activation via COPII-coated vesicles. While COPII recognizes SREBP through its escort protein SCAP, factor(s) specifically promoting SREBP/SCAP loading to the COPII machinery remains unknown. Here, we show that the ER/lipid droplet-associated protein Cideb selectively promotes the loading of SREBP/SCAP into COPII vesicles. Sterol deprivation releases SCAP from Insig and enhances ER export of SREBP/SCAP by inducing SCAP-Cideb interaction, thereby modulating sterol sensitivity. Moreover, Cideb binds to the guanine nucleotide exchange factor Sec12 to enrich SCAP/SREBP at ER exit sites, where assembling of COPII complex initiates. Loss of Cideb inhibits the cargo loading of SREBP/SCAP, reduces SREBP activation, and alleviates diet-induced hepatic steatosis. Our data point to a linchpin role of Cideb in regulated ER export of SREBP and lipid homeostasis.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)