Merck
  • Interaction of beta-funaltrexamine with [3H]cycloFOXY binding in rat brain: further evidence that beta-FNA alkylates the opioid receptor complex.
Interaction of beta-funaltrexamine with [3H]cycloFOXY binding in rat brain: further evidence that beta-FNA alkylates the opioid receptor complex.
Synapse (New York, N.Y.) (1991-06-01)
R B Rothman, V Bykov, A Mahboubi, J B Long, Q Jiang, F Porreca, B R de Costa, A E Jacobson, K C Rice, J W Holaday
ABSTRACT
beta-Funaltrexamine (beta-FNA) is an alkylating derivative of naltrexone. In addition to acting as an irreversible inhibitor of mu-receptor-mediated physiological effects, intracerebroventricular (i.c.v.) administration of beta-FNA to rat attenuates the ability of selective delta receptor antagonists and naloxone to reverse delta receptor-mediated effects. Moreover, recent work demonstrated that i.c.v. administration of beta-FNA alters the conformation of the opioid receptor complex, as inferred by a decrease in the Bmax of the lower affinity [3H][D-ala2,D-leu5]enkephalin binding site. Consistent with the decreased potency of naloxone as an inhibitor of delta receptor mediated effects, beta-FNA doubled the naloxone IC50 for displacing [3H][D-ala2,D-leu5]enkephalin from its lower affinity binding site. These data collectively support the hypothesis that the opioid receptor complex postulated to mediate mu-delta interactions in vivo is identical to the opioid receptor complex as defined by vitro ligand binding studies. A direct prediction of this hypothesis is that beta-FNA should increase the Kd of antagonists for the mu binding site (mu cx) of the receptor complex. The data reported in this paper demonstrate that beta-FNA doubled the IC50 of the potent narcotic antagonist, 6-desoxy-6 beta-fluoronaltrexone (cycloFOXY) for displacing [3H][D-ala2,D-leu5]enkephalin from its lower affinity binding site, and doubled the Kd of [3H]cycloFOXY for its mu binding site, providing additional data that the mu binding site labeled by [3H]cycloFOXY is the mu binding site of the opioid receptor complex. beta-FNA also altered the kappa binding site labeled by [3H]cycloFOXY, and when administered intrathecally to mice, beta-FNA produced a longlasting antinociception in the acetic acid writhing test.