Merck
DNA sequencing by primer walking with strings of contiguous hexamers.
Science (New York, N.Y.) (1992-12-11)
J Kieleczawa, J J Dunn, F W Studier
ABSTRACT
When template DNA is saturated with a single-stranded DNA binding protein (SSB), strings of three or four contiguous hexanucleotides (hexamers) can cooperate through base-stacking interactions to prime DNA synthesis specifically from the 3' end of the string. Under the same conditions, priming by individual hexamers is suppressed. Strings of three of four hexamers representing more than 200 of the 4096 possible hexamers primed easily readable sequence ladders at more than 75 different sites in single-stranded or denatured double-stranded templates 6.4 kilobases to 40 kilobase pairs long, with a success rate of 60 to 90 percent. A synthesis of 1 micromole of hexamer supplies enough material for thousands of primings, so multiple libraries of all 4096 hexamers could be distributed at a reasonable cost. Such libraries would allow rapid and economical sequencing. Automating this strategy could increase the speed and efficiency of large-scale DNA sequencing by at least an order of magnitude.
Sigma-Aldrich
Single-strand Binding Protein from Escherichia coli, >95% (SDS-PAGE), Single strand binding protein to enhance PCR specificity