Merck
  • Family-wide characterization of the DENN domain Rab GDP-GTP exchange factors.
Family-wide characterization of the DENN domain Rab GDP-GTP exchange factors.
The Journal of cell biology (2010-10-13)
Shin-ichiro Yoshimura, Andreas Gerondopoulos, Andrea Linford, Daniel J Rigden, Francis A Barr
ABSTRACT
A key requirement for Rab function in membrane trafficking is site-specific activation by GDP-GTP exchange factors (GEFs), but the majority of the 63 human Rabs have no known GEF. We have performed a systematic characterization of the 17 human DENN domain proteins and demonstrated that they are specific GEFs for 10 Rabs. DENND1A/1B localize to clathrin patches at the plasma membrane and activate Rab35 in an endocytic pathway trafficking Shiga toxin to the trans-Golgi network. DENND2 GEFs target to actin filaments and control Rab9-dependent trafficking of mannose-6-phosphate receptor to lysosomes. DENND4 GEFs target to a tubular membrane compartment adjacent to the Golgi, where they activate Rab10, which suggests a function in basolateral polarized sorting in epithelial cells that compliments the non-DENN GEF Sec2 acting on Rab8 in apical sorting. DENND1C, DENND3, DENND5A/5B, MTMR5/13, and MADD activate Rab13, Rab12, Rab39, Rab28, and Rab27A/27B, respectively. Together, these findings provide a basis for future studies on Rab regulation and function.
Sigma-Aldrich
Uridine 5′-diphosphate disodium salt hydrate, ≥96.0% (HPLC)
Sigma-Aldrich
Uridine 5′-triphosphate trisodium salt dihydrate, ≥80%