Merck
  • Distribution of AMPA receptor subunits and TARPs in synaptic and extrasynaptic membranes of the adult rat nucleus accumbens.
Distribution of AMPA receptor subunits and TARPs in synaptic and extrasynaptic membranes of the adult rat nucleus accumbens.
Neuroscience letters (2010-12-25)
Carrie R Ferrario, Jessica A Loweth, Mike Milovanovic, Xiaoting Wang, Marina E Wolf
ABSTRACT
We characterized the distribution of AMPA receptor (AMPAR) subunits and the transmembrane AMPA receptor regulatory proteins (TARPs) γ-2 and γ-4 in adult rat nucleus accumbens (NAc) using a method that separates plasma membranes into synaptic membrane-enriched and extrasynaptic membrane-enriched fractions. We also measured GluA1 phosphorylated at serine 845 (pS845 GluA1) and serine 831 (pS831 GluA1). GluA1-3 protein levels and pS831 GluA1/total GluA1 were higher in synaptic membranes. However, pS845 GluA1/total GluA1 was higher in extrasynaptic membranes, consistent with a role for S845 phosphorylation in GluA1 insertion at extrasynaptic sites. Homeric GluA1 receptors were detected in extrasynaptic membranes, consistent with evidence for extrasynaptic Ca(2+)-permeable AMPARs in other systems. The TARP γ-2 was enriched in synaptic membranes, whereas γ-4 was mainly found in extrasynaptic membranes, suggesting distinct roles for these proteins in the NAc. These experiments provide fundamental information that will aid in the interpretation of studies on AMPAR-related plasticity in the NAc.
Sigma-Aldrich
Anti-Glutamate Receptor 2 & 3 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Calcium Channel γ 4 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Anti-NMDAR 2B (984-1104) Rabbit pAb, lyophilized, Calbiochem®