Merck
  • Regulation of p53 by Mdm2 E3 ligase function is dispensable in embryogenesis and development, but essential in response to DNA damage.
Regulation of p53 by Mdm2 E3 ligase function is dispensable in embryogenesis and development, but essential in response to DNA damage.
Cancer cell (2014-08-15)
Laura A Tollini, Aiwen Jin, Jikyoung Park, Yanping Zhang
ABSTRACT
Mdm2 E3 ubiquitin ligase-mediated p53 degradation is generally accepted as the major mechanism for p53 regulation; nevertheless, the in vivo significance of this function has not been unequivocally established. Here, we have generated an Mdm2(Y487A) knockin mouse; Mdm2(Y487A) mutation inactivates Mdm2 E3 ligase function without affecting its ability to bind its homolog MdmX. Unexpectedly, Mdm2(Y487A/Y487A) mice were viable and developed normally into adulthood. While disruption of Mdm2 E3 ligase function resulted in p53 accumulation, p53 transcriptional activity remained low; however, exposure to sublethal stress resulted in hyperactive p53 and p53-dependent mortality in Mdm2(Y487A/Y487A) mice. These findings reveal a potentially dispensable nature for Mdm2 E3 ligase function in p53 regulation, providing insight that may affect how this pathway is targeted therapeutically.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
1 Available to ship on January 03, 2025
Details...
$151.00
$135.90
25 mg
1 Available to ship on January 03, 2025
Details...
$479.00