Merck
TFAP2 transcription factors are regulators of lipid droplet biogenesis.
eLife (2018-09-27)
Cameron C Scott, Stefania Vossio, Jacques Rougemont, Jean Gruenberg
ABSTRACT
How trafficking pathways and organelle abundance adapt in response to metabolic and physiological changes is still mysterious, although a few transcriptional regulators of organellar biogenesis have been identified in recent years. We previously found that the Wnt signaling directly controls lipid droplet formation, linking the cell storage capacity to the established functions of Wnt in development and differentiation. In the present paper, we report that Wnt-induced lipid droplet biogenesis does not depend on the canonical TCF/LEF transcription factors. Instead, we find that TFAP2 family members mediate the pro-lipid droplet signal induced by Wnt3a, leading to the notion that the TFAP2 transcription factor may function as a 'master' regulator of lipid droplet biogenesis.
Sigma-Aldrich
MISSION® esiRNA, targeting human DGAT2
Sigma-Aldrich
MISSION® esiRNA, targeting human DDIT3
Sigma-Aldrich
MISSION® esiRNA, targeting human DGAT1
Sigma-Aldrich
MISSION® esiRNA, targeting human SOAT1
Sigma-Aldrich
MISSION® esiRNA, targeting human TFAP2C
Sigma-Aldrich
MISSION® esiRNA, targeting human TFAP2A
Sigma-Aldrich
MISSION® esiRNA, targeting human GSK3B