Merck
  • The Reactive Oxygen Species-Mitophagy Signaling Pathway Regulates Liver Endothelial Cell Survival During Ischemia/Reperfusion Injury.
The Reactive Oxygen Species-Mitophagy Signaling Pathway Regulates Liver Endothelial Cell Survival During Ischemia/Reperfusion Injury.
Liver transplantation : official publication of the American Association for the Study of Liver Diseases and the International Liver Transplantation Society (2018-07-25)
Ricky H Bhogal, Christopher J Weston, Susanne Velduis, Henri G D Leuvenink, Gary M Reynolds, Scott Davies, Luu Nyguet-Thin, Mohammed Alfaifi, Emma L Shepard, Yuri Boteon, Lorraine Wallace, Ye H Oo, David H Adams, Darius F Mirza, Hynek Mergental, Gillian Muirhead, Barnaby T F Stephenson, Simon C Afford
ABSTRACT
Ischemia/reperfusion injury (IRI) is the main cause of complications following liver transplantation. Reactive oxygen species (ROS) were thought to be the main regulators of IRI. However, recent studies demonstrate that ROS activate the cytoprotective mechanism of autophagy promoting cell survival. Liver IRI initially damages the liver endothelial cells (LEC), but whether ROS-autophagy promotes cell survival in LEC during IRI is not known. Primary human LEC were isolated from human liver tissue and exposed to an in vitro model of IRI to assess the role of autophagy in LEC. The role of autophagy during liver IRI in vivo was assessed using a murine model of partial liver IRI. During IRI, ROS specifically activate autophagy-related protein (ATG) 7 promoting autophagic flux and the formation of LC3B-positive puncta around mitochondria in primary human LEC. Inhibition of ROS reduces autophagic flux in LEC during IRI inducing necrosis. In addition, small interfering RNA knockdown of ATG7 sensitized LEC to necrosis during IRI. In vivo murine livers in uninjured liver lobes demonstrate autophagy within LEC that is reduced following IRI with concomitant reduction in autophagic flux and increased cell death. In conclusion, these findings demonstrate that during liver IRI ROS-dependent autophagy promotes the survival of LEC, and therapeutic targeting of this signaling pathway may reduce liver IRI following transplantation.
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
2′,7′-Dichlorofluorescin Diacetate, Cell-permeable fluorogenic probe that is useful for the detection of reactive oxygen species (ROS) and nitric oxide (•NO) and for the determination of the degree of overall oxidative stress.
Sigma-Aldrich
MISSION® esiRNA, targeting human ATG7
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trizma® hydrochloride, reagent grade, ≥99.0% (titration), crystalline
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)