Merck
  • GeH: a novel material as a visible-light driven photocatalyst for hydrogen evolution.
GeH: a novel material as a visible-light driven photocatalyst for hydrogen evolution.
Chemical communications (Cambridge, England) (2014-08-08)
Zhenhua Liu, Zaizhu Lou, Zhujie Li, Gang Wang, Zeyan Wang, Yuanyuan Liu, Baibiao Huang, Shengqing Xia, Xiaoyan Qin, Xiaoyang Zhang, Ying Dai
ABSTRACT
A novel 2D material germanane (GeH), which was synthesized by an ion-exchange approach, was firstly found to exhibit photocatalytic performance in the hydrogen evolution of water splitting and decomposition of organic contaminants under illumination of visible light (λ ≥ 420 nm).
Sigma-Aldrich
Methyl germanane
Pricing and availability is not currently available.
Sigma-Aldrich
Allyl germanane
Pricing and availability is not currently available.
Sigma-Aldrich
Germanane
Pricing and availability is not currently available.