Merck
  • Cysteine-Independent Inhibition of Alzheimer's Disease-like Paired Helical Filament Assembly by Leuco-Methylthioninium (LMT).
Cysteine-Independent Inhibition of Alzheimer's Disease-like Paired Helical Filament Assembly by Leuco-Methylthioninium (LMT).
Journal of molecular biology (2018-08-20)
Youssra K Al-Hilaly, Saskia J Pollack, Janet E Rickard, Michael Simpson, Ana-Caroline Raulin, Thomas Baddeley, Pascale Schellenberger, John M D Storey, Charles R Harrington, Claude M Wischik, Louise C Serpell
ABSTRACT
Alzheimer's disease is a tauopathy characterized by pathological fibrillization of tau protein to form the paired helical filaments (PHFs), which constitute neurofibrillary tangles. The methylthioninium (MT) moiety reverses the proteolytic stability of the PHF core and is in clinical development for treatment of Alzheimer's disease in a stable reduced form as leuco-MT. It has been hypothesized that MT acts via oxidation of cysteine residues, which is incompatible with activity in the predominantly reducing environment of living cells. We have shown recently that the PHF-core tau unit assembles spontaneously in vitro to form PHF-like filaments. Here we describe studies using circular dichroism, SDS-PAGE, transmission electron microscopy and site-directed mutagenesis to elucidate the mechanism of action of the MT moiety. We show that MT inhibitory activity is optimal in reducing conditions, that the active moiety is the reduced leuco-MT form of the molecule and that its mechanism of action is cysteine independent.
Product Description
Methylthioninium chloride, European Pharmacopoeia (EP) Reference Standard