Merck
  • o-Carboranylalkoxy-1,3,5-Triazine Derivatives: Synthesis, Characterization, X-ray Structural Studies, and Biological Activity.
o-Carboranylalkoxy-1,3,5-Triazine Derivatives: Synthesis, Characterization, X-ray Structural Studies, and Biological Activity.
Molecules (Basel, Switzerland) (2018-09-12)
Guo Fan Jin, Hyun Seung Ban, Hiroyuki Nakamura, Jong-Dae Lee
ABSTRACT
Morpholine- and bis(2-methoxyethyl)amine-substituted 1,3,5-triazine derivatives containing an alkoxy-o-carborane in the 6-position of the triazine ring were successfully synthesized. The molecular structures of the methoxy- and ethoxy-o-carboranyl-1,3,5-triazines were established by X-ray crystallography. In vitro studies showed that the methylene bridged morpholine- and bis(2-methoxyethyl)amine-substituted o-carboranyl-1,3,5-triazines accumulated to high levels in B16 melanoma cells and exhibited higher cytotoxicity than p-boronophenylalanine.
Sigma-Aldrich
2-Butyne-1,4-diol, 99%
Sigma-Aldrich
3-Butyn-1-ol, 97%
Sigma-Aldrich
Propargyl alcohol, 99%
Sigma-Aldrich
4-Pentyn-1-ol, 97%
Sigma-Aldrich
Bis(2-methoxyethyl)amine, 98%